Anti-SULT2A1 Mouse Monoclonal Antibody [clone: 4D7]

Lieferant: OriGene
ORIGCF501694EA 749 EUR
ORIGCF501694
Anti-SULT2A1 Mouse Monoclonal Antibody [clone: 4D7]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT2A1
Clonality: Monoclonal
Clone: 4D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR