Anti-RIT2 Mouse Monoclonal Antibody [clone: 4B5]

Lieferant: OriGene
ORIGCF501704EA 749 EUR
ORIGCF501704
Anti-RIT2 Mouse Monoclonal Antibody [clone: 4B5]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RIT2
Clonality: Monoclonal
Clone: 4B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR