Anti-CD80 Mouse Monoclonal Antibody [clone: 7H3]

Lieferant: OriGene
ORIGCF501669EA 749 EUR
ORIGCF501669
Anti-CD80 Mouse Monoclonal Antibody [clone: 7H3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 7H3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR