Anti-HP Mouse Monoclonal Antibody [clone: 1F9]

Lieferant: OriGene
ORIGCF501674EA 749 EUR
ORIGCF501674
Anti-HP Mouse Monoclonal Antibody [clone: 1F9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HP
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR