Anti-KCTD14 Mouse Monoclonal Antibody [clone: 2B9]

Lieferant: OriGene
ORIGCF502662EA 770 EUR
ORIGCF502662
Anti-KCTD14 Mouse Monoclonal Antibody [clone: 2B9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KCTD14
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR