Anti-NPR3 Mouse Monoclonal Antibody [clone: 11A6]

Lieferant: OriGene
ORIGCF501076EA 749 EUR
ORIGCF501076
Anti-NPR3 Mouse Monoclonal Antibody [clone: 11A6]
Antikörper
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: NPR3
Clonality: Monoclonal
Clone: 11A6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR