Anti-SHC1 Mouse Monoclonal Antibody [clone: 3H1]

Lieferant: OriGene
ORIGCF501103EA 749 EUR
ORIGCF501103
Anti-SHC1 Mouse Monoclonal Antibody [clone: 3H1]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: SHC1
Clonality: Monoclonal
Clone: 3H1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR