Anti-UHMK1 Mouse Monoclonal Antibody [clone: 2D3]

Lieferant: OriGene
ORIGCF501719EA 749 EUR
ORIGCF501719
Anti-UHMK1 Mouse Monoclonal Antibody [clone: 2D3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: UHMK1
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR