Anti-ERCC1 Mouse Monoclonal Antibody [clone: 2A9]

Lieferant: OriGene
ORIGCF501726EA 764 EUR
ORIGCF501726
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 2A9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR