Anti-CNDP1 Mouse Monoclonal Antibody [clone: 2D2]

Lieferant: OriGene
ORIGCF502093EA 749 EUR
ORIGCF502093
Anti-CNDP1 Mouse Monoclonal Antibody [clone: 2D2]
Antikörper
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: CNDP1
Clonality: Monoclonal
Clone: 2D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR