Anti-MYD88 Mouse Monoclonal Antibody [clone: 1B4]

Lieferant: OriGene
ORIGCF502104EA 770 EUR
ORIGCF502104
Anti-MYD88 Mouse Monoclonal Antibody [clone: 1B4]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MYD88
Clonality: Monoclonal
Clone: 1B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR