Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 4E2]

Lieferant: OriGene
ORIGCF502130EA 749 EUR
ORIGCF502130
Anti-SQSTM1 Mouse Monoclonal Antibody [clone: 4E2]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SQSTM1
Clonality: Monoclonal
Clone: 4E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR