Anti-NDUFA7 Mouse Monoclonal Antibody [clone: 2C3]

Lieferant: OriGene
ORIGCF502761EA 749 EUR
ORIGCF502761
Anti-NDUFA7 Mouse Monoclonal Antibody [clone: 2C3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NDUFA7
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR