Anti-FGF21 Mouse Monoclonal Antibody [clone: 4B2]

Lieferant: OriGene
ORIGCF502771EA 770 EUR
ORIGCF502771
Anti-FGF21 Mouse Monoclonal Antibody [clone: 4B2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FGF21
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR