Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 15E2]

Lieferant: OriGene
ORIGCF501156EA 749 EUR
ORIGCF501156
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 15E2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: CYP1A2
Clonality: Monoclonal
Clone: 15E2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR