Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 7A9]

Lieferant: OriGene
ORIGCF501157EA 749 EUR
ORIGCF501157
Anti-CYP1A2 Mouse Monoclonal Antibody [clone: 7A9]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: CYP1A2
Clonality: Monoclonal
Clone: 7A9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR