Anti-TUBA8 Mouse Monoclonal Antibody [clone: 3G2]

Lieferant: OriGene
ORIGCF501159EA 749 EUR
ORIGCF501159
Anti-TUBA8 Mouse Monoclonal Antibody [clone: 3G2]
Antikörper
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TUBA8
Clonality: Monoclonal
Clone: 3G2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR