Anti-GSTA4 Mouse Monoclonal Antibody [clone: 1E2]

Lieferant: OriGene
ORIGCF501752EA 749 EUR
ORIGCF501752
Anti-GSTA4 Mouse Monoclonal Antibody [clone: 1E2]
Antikörper
Recommended Dilutions: Western Blot: 1:200-1000

Type: Primary
Antigen: GSTA4
Clonality: Monoclonal
Clone: 1E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR