Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: 7G9]

Lieferant: OriGene
ORIGCF501762EA 770 EUR
ORIGCF501762
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: 7G9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PIK3AP1
Clonality: Monoclonal
Clone: 7G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR