Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1A8]

Lieferant: OriGene
ORIGCF501767EA 749 EUR
ORIGCF501767
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1A8]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR