Anti-GSTT2 Mouse Monoclonal Antibody [clone: 6B7]

Lieferant: OriGene
ORIGCF501830EA 770 EUR
ORIGCF501830
Anti-GSTT2 Mouse Monoclonal Antibody [clone: 6B7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: GSTT2
Clonality: Monoclonal
Clone: 6B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR