Anti-LMAN1 Mouse Monoclonal Antibody [clone: 1A8]

Lieferant: OriGene
ORIGCF502137EA 749 EUR
ORIGCF502137
Anti-LMAN1 Mouse Monoclonal Antibody [clone: 1A8]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LMAN1
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR