Anti-AFP Mouse Monoclonal Antibody [clone: 2H7]

Lieferant: OriGene
ORIGCF502169EA 749 EUR
ORIGCF502169
Anti-AFP Mouse Monoclonal Antibody [clone: 2H7]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: 2H7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR