Anti-ALDH1A3 Mouse Monoclonal Antibody [clone: 4B4]

Lieferant: OriGene
ORIGCF502842EA 749 EUR
ORIGCF502842
Anti-ALDH1A3 Mouse Monoclonal Antibody [clone: 4B4]
Antikörper
Recommended Dilutions: Western Blot: 1:200-2000; Immunochemistry: 1:150

Type: Primary
Antigen: ALDH1A3
Clonality: Monoclonal
Clone: 4B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR