Anti-PPM1B Mouse Monoclonal Antibody [clone: 3C9]

Lieferant: OriGene
ORIGCF502866EA 749 EUR
ORIGCF502866
Anti-PPM1B Mouse Monoclonal Antibody [clone: 3C9]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPM1B
Clonality: Monoclonal
Clone: 3C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR