Anti-ERCC1 Mouse Monoclonal Antibody [clone: 2A7]

Lieferant: OriGene
ORIGCF501182EA 749 EUR
ORIGCF501182
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 2A7]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR