Anti-BAT1 Mouse Monoclonal Antibody [clone: 2C5]

Lieferant: OriGene
ORIGCF501186EA 749 EUR
ORIGCF501186
Anti-BAT1 Mouse Monoclonal Antibody [clone: 2C5]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: BAT1
Clonality: Monoclonal
Clone: 2C5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR