Anti-Stat4 Mouse Monoclonal Antibody [clone: 2A2]

Lieferant: OriGene
ORIGCF500005EA 770 EUR
ORIGCF500005
Anti-Stat4 Mouse Monoclonal Antibody [clone: 2A2]
Antikörper
Recommended Dilutions:Western Blot: 1:100; Immunochemistry: 1:50

Type: Primary
Antigen: Stat4
Clonality: Monoclonal
Clone: 2A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR