Anti-HHex Mouse Monoclonal Antibody [clone: 3C4]

Lieferant: OriGene
ORIGCF500026EA 749 EUR
ORIGCF500026
Anti-HHex Mouse Monoclonal Antibody [clone: 3C4]
Antikörper
Recommended Dilutions:Western Blot: 1:50-1:100; Immunofluorescence: 1:100

Type: Primary
Antigen: HHex
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR