Anti-NTF4 Mouse Monoclonal Antibody [clone: 4D2]

Lieferant: OriGene
ORIGCF500033EA 749 EUR
ORIGCF500033
Anti-NTF4 Mouse Monoclonal Antibody [clone: 4D2]
Antikörper
Recommended Dilutions: Western Blot: 1:20000

Type: Primary
Antigen: NTF4
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR