Anti-HDAC6 Mouse Monoclonal Antibody [clone: 3E7]

Lieferant: OriGene
ORIGCF502177EA 749 EUR
ORIGCF502177
Anti-HDAC6 Mouse Monoclonal Antibody [clone: 3E7]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: HDAC6
Clonality: Monoclonal
Clone: 3E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR