Anti-HDAC1 Mouse Monoclonal Antibody [clone: 5F9]

Lieferant: OriGene
ORIGCF502193EA 749 EUR
ORIGCF502193
Anti-HDAC1 Mouse Monoclonal Antibody [clone: 5F9]
Antikörper
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: HDAC1
Clonality: Monoclonal
Clone: 5F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR