Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 1E9]

Lieferant: OriGene
ORIGCF502194EA 749 EUR
ORIGCF502194
Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 1E9]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: DNAJB1
Clonality: Monoclonal
Clone: 1E9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR