Anti-MTMR2 Mouse Monoclonal Antibody [clone: 5G5]

Lieferant: OriGene
ORIGCF502204EA 749 EUR
ORIGCF502204
Anti-MTMR2 Mouse Monoclonal Antibody [clone: 5G5]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: MTMR2
Clonality: Monoclonal
Clone: 5G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR