Anti-CRYAB Mouse Monoclonal Antibody [clone: 4A6]

Lieferant: OriGene
ORIGCF500582EA 749 EUR
ORIGCF500582
Anti-CRYAB Mouse Monoclonal Antibody [clone: 4A6]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 4A6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR