Anti-PRKY Mouse Monoclonal Antibody [clone: 3E10]

Lieferant: OriGene
ORIGCF500569EA 749 EUR
ORIGCF500569
Anti-PRKY Mouse Monoclonal Antibody [clone: 3E10]
Antikörper
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PRKY
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR