Anti-SLC2A5 Mouse Monoclonal Antibody [clone: 11E1]

Lieferant: OriGene
ORIGCF500574EA 749 EUR
ORIGCF500574
Anti-SLC2A5 Mouse Monoclonal Antibody [clone: 11E1]
Antikörper
Recommended Dilutions: Western Blot: 1:5000; Immunofluorescence: 1:100

Type: Primary
Antigen: SLC2A5
Clonality: Monoclonal
Clone: 11E1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR