Anti-CRYAB Mouse Monoclonal Antibody [clone: 3E3]

Lieferant: OriGene
ORIGCF500580EA 749 EUR
ORIGCF500580
Anti-CRYAB Mouse Monoclonal Antibody [clone: 3E3]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR