Anti-BIRC5 Mouse Monoclonal Antibody [clone: 2D9]

Lieferant: OriGene
ORIGCF501245EA 749 EUR
ORIGCF501245
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 2D9]
Antikörper
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BIRC5
Clonality: Monoclonal
Clone: 2D9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR