Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 2H3]

Lieferant: OriGene
ORIGCF500049EA 749 EUR
ORIGCF500049
Anti-CTNNB1 Mouse Monoclonal Antibody [clone: 2H3]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000

Type: Primary
Antigen: CTNNB1
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR