Anti-CPA1 Mouse Monoclonal Antibody [clone: 2A3]

Lieferant: OriGene
ORIGCF500053EA 770 EUR
ORIGCF500053
Anti-CPA1 Mouse Monoclonal Antibody [clone: 2A3]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: CPA1
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR