Anti-NMNAT1 Mouse Monoclonal Antibody [clone: 1G4]

Lieferant: OriGene
ORIGCF502220EA 749 EUR
ORIGCF502220
Anti-NMNAT1 Mouse Monoclonal Antibody [clone: 1G4]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NMNAT1
Clonality: Monoclonal
Clone: 1G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR