Anti-CDCP1 Mouse Monoclonal Antibody [clone: 5B3]

Lieferant: OriGene
ORIGCF502231EA 749 EUR
ORIGCF502231
Anti-CDCP1 Mouse Monoclonal Antibody [clone: 5B3]
Antikörper
Recommended Dilutions: Western Blot: 1:500-1000; Flow Cytometry: 1:100

Type: Primary
Antigen: CDCP1
Clonality: Monoclonal
Clone: 5B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR