Anti-LMAN1 Mouse Monoclonal Antibody [clone: 1C5]

Lieferant: OriGene
ORIGCF502237EA 749 EUR
ORIGCF502237
Anti-LMAN1 Mouse Monoclonal Antibody [clone: 1C5]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: LMAN1
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR