Anti-GBP5 Mouse Monoclonal Antibody [clone: 5C9]

Lieferant: OriGene
ORIGCF502242EA 770 EUR
ORIGCF502242
Anti-GBP5 Mouse Monoclonal Antibody [clone: 5C9]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GBP5
Clonality: Monoclonal
Clone: 5C9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR