Anti-PPP5C Mouse Monoclonal Antibody [clone: 3G1]

Lieferant: OriGene
ORIGCF500597EA 749 EUR
ORIGCF500597
Anti-PPP5C Mouse Monoclonal Antibody [clone: 3G1]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPP5C
Clonality: Monoclonal
Clone: 3G1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR