Anti-MAD2L1 Mouse Monoclonal Antibody [clone: 4D2]

Lieferant: OriGene
ORIGCF500617EA 749 EUR
ORIGCF500617
Anti-MAD2L1 Mouse Monoclonal Antibody [clone: 4D2]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAD2L1
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR