Anti-SNX9 Mouse Monoclonal Antibody [clone: 1B5]

Lieferant: OriGene
ORIGCF501251EA 749 EUR
ORIGCF501251
Anti-SNX9 Mouse Monoclonal Antibody [clone: 1B5]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNX9
Clonality: Monoclonal
Clone: 1B5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR