Anti-TACC3 Mouse Monoclonal Antibody [clone: 1E8]

Lieferant: OriGene
ORIGCF501266EA 749 EUR
ORIGCF501266
Anti-TACC3 Mouse Monoclonal Antibody [clone: 1E8]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: TACC3
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR