Anti-FHL1 Mouse Monoclonal Antibody [clone: 2E11]

Lieferant: OriGene
ORIGCF501279EA 749 EUR
ORIGCF501279
Anti-FHL1 Mouse Monoclonal Antibody [clone: 2E11]
Antikörper
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: FHL1
Clonality: Monoclonal
Clone: 2E11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR